ContactUsLinkComponent
N,N-Dimethylformamide
  86007.980
 :  
undefined
N,N-Dimethylformamide
  86007.980
 :  
  • Pk:
    950 L
  • Seq:
    5783
 :
 :73.09 g/mol
 :13,03269
 :153 °C (1013 hPa)
 :–61 °C
 :0.949 g/cm³ (20 °C)
 :57.5 °C (closed cup)
 :Ambient
 :200-679-5
 :2265
 :3,III
 :01-2119475605-32
 :GHS02
 :GHS08
 :GHS07
 :68-12-2
 :HCON(CH₃)₂
 :MFCD00003284