ContactUsLinkComponent
VWR® Safe-Tamper System
VWR® Safe-Tamper System
Catalog # 720-1985
Supplier:  VWR Collection
undefined
VWR® Safe-Tamper System
Catalog # 720-1985
Supplier:  VWR Collection
Specifications
  • Description:
    Safe-Tamper System anti-tampering bag
  • L×W:
    157×306 mm
  • Pk:
    500
Specifications
About this item
Anti-tamper disposable bags, in PE, with double pocket for internal hospital biological sample transportation. Double pocket bags: Two separate compartments for lab document and specimen. It is possible to customise the bags.
  • Transparent bags, yellow coloured to identify leakage of fluid and for a better visibility of the contents
  • Adhesive closure to prevent tampering and to preserve patient privacy
  • Printed biohazard logo and instructions
  • The bag is destroyed by opening it, preventing reuse and avoiding any risk of cross-contamination