Dishes, crystallising
Supplier: Kavalier
Certificates
About this item
Borosilicate glass 3,3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For a wide range of applications.
Frequently Bought Together