

Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide) 98%
:82911-69-1
:
:C₁₉H₁₅NO₅
:MFCD00010733
:337.3 g/mol
:145…153 °C
:Refrigerator
:433-520-5
:3077
:9,III
:GHS07
:GHS09
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:145 to 153 °C
- HPLC:≥97.5 %
Frequently Bought Together