(R)-(+)-2-Methyl-CBS-oxazaborolidine 1 mol/l in toluene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:112022-83-0
:
Formula:C₁₈H₂₀BNO
MDL Number:MFCD00078440
:277.17 g/mol
:M-Clarity™ quality level = MQ200
Frequently Bought Together