Order Entry
Czech Republic
ContactUsLinkComponent
Benzo[a]pyrene-[D₁₂] (98% D), Sigma-Aldrich®
Health
Environment
Benzo[a]pyrene-[D₁₂] (98% D), Sigma-Aldrich®
Catalog # SIAL451797-10MG
Supplier:  Merck
CAS Number:  
Health
Environment
Benzo[a]pyrene-[D₁₂] (98% D), Sigma-Aldrich®
Catalog # SIAL451797-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
 :GHS08
 :GHS09
CAS Number:63466-71-7
Formula:C₂₀D₁₂