Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Benzo[a]pyrene-[D12]
Benzo[a]pyrene-[D₁₂]
Benzo[a]pyrene-[D₁₂]
Health
Environment
Benzo[a]pyrene-[D₁₂]
Catalog # EHERC20635100
Supplier:
EHRENSTORFER
CAS Number:
Expand image
Health
Environment
Benzo[a]pyrene-[D₁₂]
Catalog # EHERC20635100
Supplier:
EHRENSTORFER
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
10 mg
Seq:
2275
Specifications
About this item
Product Details
:
:
GHS08
:
GHS09
CAS Number:
63466-71-7
Formula:
C₂₀D₁₂
Product Details