

Bis(1,5-cyclooctadiene)rhodium(I) hexafluoroantimonate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:130296-28-5
:
Formula:C₁₆H₂₄F₆RhSb
MDL Number:MFCD11113031
:555.01 g/mol
ADR:9,III
:GHS07
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together