Chloro(1,5-cyclooctadiene)rhodium(I) dimer
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12092-47-6
:
Formula:C₁₆H₂₄Cl₂Rh₂
MDL Number:MFCD00012415
:493.08 g/mol
Melting Pt:243 °C (dec.)
Storage Temperature:Ambient
EINECS:235-157-6
Related Information
Frequently Bought Together