Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Chloro(1,5-cyclooctadiene)iridium (I) dimer
Chloro(1,5-cyclooctadiene)iridium(I) dimer ≥99%
Chloro(1,5-cyclooctadiene)iridium(I) dimer ≥99%
Chloro(1,5-cyclooctadiene)iridium(I) dimer ≥99%
ABCR77-0400.0500
:
abcr
:
Chloro(1,5-cyclooctadiene)iridium(I) dimer ≥99%
ABCR77-0400.0500
:
abcr
:
Pack type:
Glass bottle for solids
Pk:
500 mg
:
:
671.70 g/mol
:
190 °C (dec.)
:
235-170-7
:
12112-67-3
:
C₁₆H₂₄Cl₂Ir₂
:
MFCD00012414