Order Entry
Czech Republic
ContactUsLinkComponent
 
Benzo[a]pyrene-[D12] 100 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERXA20635100CY
 :  
Benzo[a]pyrene-[D12] 100 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERXA20635100CY
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    63466-71-7
  • Description:
    Benzo[a]pyrene-[D12] 100 µg/ml in cyclohexane
  • Formula:
    C₂₀D₁₂
  • Description:
    Benzo[a]pyrene-[D12]