
(Bis(trifluoroacetoxy)iodo)benzene 98%
Catalog # ACRO213670100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:121 to 125 °C
- Titration Iodometric:≥97.5 %
Specification Test Results
About this item
:
:430.04 g/mol
Melting Pt:121…125 °C
EINECS:220-308-0
:GHS07
Storage Temperature:Ambient
CAS Number:2712-78-9
Formula:C₁₀H₅F₆IO₄
MDL Number:MFCD00009672