β-Bromostyrene (mixture of cis and trans isomers), Sigma-Aldrich®
Catalog # SIAL157449-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:183.05 g/mol
:M-Clarity™ quality level = MQ100
EINECS:203-131-3
CAS Number:103-64-0
Formula:C₈H₇Br
MDL Number:MFCD00000185