
7-Hydroxyflavanone 98%
Catalog # ACRO364170010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:189 to 197 °C
- HPLC:≥97.5 %
Specification Test Results
About this item
:
:240.25 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:6515-36-2
Formula:C₁₅H₁₂O₃
MDL Number:MFCD00017487