Order Entry
Czech Republic
ContactUsLinkComponent
Cyanogen bromide 3 M in dichloromethane
Toxic
Health
Corrosive
Environment
Cyanogen bromide 3 M in dichloromethane
Catalog # ACRO353991000
CAS Number:  
undefined
Toxic
Health
Corrosive
Environment
Cyanogen bromide 3 M in dichloromethane
Catalog # ACRO353991000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Solution
  • Strength:
    2.95 to 3.3 M

Specification Test Results

About this item

 :
 :105.92 g/mol
Storage Temperature:Refrigerator
EINECS:208-051-2
UN:2927
ADR:6.1,I
 :GHS06
 :GHS08
 :GHS05
 :GHS09
CAS Number:506-68-3
Formula:CBrN
MDL Number:MFCD00011597