
cis-4-Aminocyclohexanecarboxylic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3685-23-2
:
Formula:C₇H₁₃NO₂
MDL Number:MFCD00191730
:143.19 g/mol
Melting Pt:299…301 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together