Order Entry
Czech Republic
ContactUsLinkComponent
cis-4-Aminocyclohexanecarboxylic acid
cis-4-Aminocyclohexanecarboxylic acid
Catalog # ABCRAB135321.0005
Supplier:  abcr
CAS Number:  
cis-4-Aminocyclohexanecarboxylic acid
Catalog # ABCRAB135321.0005
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :143.19 g/mol
CAS Number:3685-23-2
Formula:C₇H₁₃NO₂
MDL Number:MFCD00191730