Diethyl allylphosphonate, Sigma-Aldrich®
Catalog # SIAL565415-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:178.1660 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:1067-87-4
Formula:C₇H₁₅O₃P
MDL Number:MFCD00015134