Order Entry
Czech Republic
ContactUsLinkComponent
 
Pesticide reference standard, Piperonyl butoxide 10 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERL16240000AL
 :  
Pesticide reference standard, Piperonyl butoxide 10 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERL16240000AL
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    51-03-6
  • Description:
    Piperonyl butoxide 10 µg/ml in acetonitrile
  • Formula:
    C₁₉H₃₀O₅
  • Description:
    Piperonyl butoxide