Order Entry
Czech Republic
ContactUsLinkComponent
 
(R)-(-)-6,6'-Dibromo-1,1'-bi-2-naphthol, Sigma-Aldrich®
Catalog # SIAL482617-500MG
Supplier:  Merck
CAS Number:  
(R)-(-)-6,6'-Dibromo-1,1'-bi-2-naphthol, Sigma-Aldrich®
Catalog # SIAL482617-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :444.12 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:65283-60-5
Formula:C₂₀H₁₂Br₂O₂
MDL Number:MFCD00798290