
(R)-(+)-1,2,3,4-Tetrahydro-1-naphthoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23357-47-3
:
Formula:C₁₁H₁₂O₂
MDL Number:MFCD07368364
:176.22 g/mol
Melting Pt:56…58 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together