Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) 98%
 
undefined
 
 
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) 98%

 

  • Appearance:
    White to beige to light brown Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    92 to 96 °C
  • Water:
    ≤0.1 % (K.F.)
  • Titration with NaOH:
    ≥97.5 %

 

 

Frequently Bought Together