4-Methyl-1-naphthoic acid, Sigma-Aldrich®
Catalog # SIAL526533-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:186.21 g/mol
CAS Number:4488-40-8
Formula:C₁₂H₁₀O₂
MDL Number:MFCD00671557