
1-Amino-4-naphthol hydrochloride tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5959-56-8
:
Formula:C₁₀H₁₀ClNO
MDL Number:MFCD00012562
Melting Pt:∼ 272 °C (dec.)
:195.65 g/mol
EINECS:227-727-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together