Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Bromo-2-naphthoic acid
1-Bromo-2-naphthoic acid, Sigma-Aldrich®
1-Bromo-2-naphthoic acid, Sigma-Aldrich®
1-Bromo-2-naphthoic acid, Sigma-Aldrich®
SIALB7252-1G
:
Merck
:
1-Bromo-2-naphthoic acid, Sigma-Aldrich®
SIALB7252-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
251.0800 g/mol
:
M-Clarity™ quality level = MQ100
:
243-984-9
:
20717-79-7
:
C₁₁H₇BrO₂
:
MFCD00021408