Order Entry
Czech Republic
ContactUsLinkComponent
8,8'-[Oxybis(ethyleneoxyethyleneoxy)]diquinoline (Kryptofix® 5) ≥99.0% for synthesis, Sigma-Aldrich®
8,8'-[Oxybis(ethyleneoxyethyleneoxy)]diquinoline (Kryptofix® 5) ≥99.0% for synthesis, Sigma-Aldrich®
Catalog # 8.11689.0005
Supplier:  Merck
CAS Number:  
8,8'-[Oxybis(ethyleneoxyethyleneoxy)]diquinoline (Kryptofix® 5) ≥99.0% for synthesis, Sigma-Aldrich®
Catalog # 8.11689.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Assay (HClO₄):
    ≥ 99.0 %
  • Melting range (lower value):
    ≥ 72 °C
  • Melting range (upper value):
    ≤ 76 °C
  • Identity (IR):
    passes test

Specification Test Results

About this item

 :
 :448.52 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:260-674-9
CAS Number:57310-75-5
Formula:C₂₆H₂₈N₂O₅
MDL Number:MFCD00040469