4-(Trifluoromethylthio)benzaldehyde, Sigma-Aldrich®
Catalog # SIAL529141-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:206.19 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:4021-50-5
Formula:C₈H₅F₃OS
MDL Number:MFCD00129191