Pesticide reference standard, 4,4'-Dibromooctafluorobiphenyl (DBOB) 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA12240600CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:10386-84-2
- Description:4,4'-Dibromooctafluorobiphenyl (DBOB) 100 µg/ml in cyclohexane
- Formula:C₁₂Br₂F₈
- Description:4,4'-Dibromooctafluorobiphenyl (DBOB)