Tetraethyl methylenebisphosphonate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1660-94-2
:
Formula:C₉H₂₂O₆P₂
MDL Number:MFCD00039887
:288.22 g/mol
:M-Clarity™ quality level = MQ200
EINECS:216-764-5
Frequently Bought Together