
Diethyl (2-bromoethyl)phosphonate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5324-30-1
:
Formula:C₆H₁₄BrO₃P
MDL Number:MFCD00000239
:245.06 g/mol
Boiling Pt:98…100 °C (1 mmHg)
Density:1.275
Flash Pt:>110 °C (230 °F)
EINECS:226-193-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together