

5-Hydroxy-1,4-naphthoquinone (juglone) 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:481-39-0
:
Formula:C₁₀H₆O₃
MDL Number:MFCD00001684
:174.16 g/mol
Melting Pt:161…163 °C
Storage Temperature:Ambient
EINECS:207-567-5
UN:2811
ADR:6.1,III
:GHS06
:GHS09
Specification Test Results
- Appearance:Orange to brown Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together