Order Entry
Czech Republic
ContactUsLinkComponent
2-Hydroxy-1,4-naphthoquinone
2-Hydroxy-1,4-naphthoquinone
Catalog # EHERC14233800
Supplier:  EHRENSTORFER
CAS Number:  
2-Hydroxy-1,4-naphthoquinone
Catalog # EHERC14233800
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :174.16 g/mol
EINECS:201-496-3
CAS Number:83-72-7
Formula:C₁₀H₆O₃
MDL Number:MFCD00001678