3,5-Bis(trifluoromethyl)phenylacetonitrile, Sigma-Aldrich®
Catalog # SIAL263427-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:253.15 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:285-294-0
UN:3276
CAS Number:85068-32-2
Formula:C₁₀H₅F₆N
MDL Number:MFCD00009904