
2-Bromo-5-methylbenzotrifluoride ≥98%
Catalog # H31523.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
CAS Number:261952-20-9
:
Formula:C₈H₆BrF₃
MDL Number:MFCD01631586
:239.04 g/mol
:GHS07
Storage Temperature:Ambient