1,3-Diphenyl-1-butanone 95%
Catalog # B20654.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:224.30 g/mol
Melting Pt:68…72 °C
Storage Temperature:Ambient
CAS Number:1533-20-6
Formula:C₁₆H₁₆O
MDL Number:MFCD00026345