
4-Bromo-2-fluoro-5-(trifluoromethyl)aniline ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:104460-70-0
:
Formula:C₇H₄BrF₄N
MDL Number:MFCD04973757
Melting Pt:30…41 °C
UN:2811
ADR:6.1,III
:258.01 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together