Organic reference standard, 2,4,5-Tribromobiphenyl (PBB No. 29) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL21002900AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:115245-07-3
- Description:2,4,5-Tribromobiphenyl (PBB No. 29) 10 µg/ml in acetonitrile
- Formula:C₁₂H₇Br₃
- Description:2,4,5-Tribromobiphenyl (PBB No. 29)