6-Fluoro-2-naphthoic acid, Sigma-Aldrich®
Catalog # SIAL638862-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:190.1700 g/mol
:M-Clarity™ quality level = MQ200
CAS Number:5043-01-6
Formula:C₁₁H₇FO₂
MDL Number:MFCD06200717