1,4-Dihydroxy-2-naphthoic acid, Sigma-Aldrich®
Catalog # SIAL281255-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:204.18 g/mol
:M-Clarity™ quality level = MQ100
EINECS:250-674-7
CAS Number:31519-22-9
Formula:C₁₁H₈O₄
MDL Number:MFCD00010370