Order Entry
Czech Republic
ContactUsLinkComponent
(4-Aminophenyl)acetonitrile 99%
Irritant
(4-Aminophenyl)acetonitrile 99%
Catalog # ACRO103210100
CAS Number:  
undefined
Irritant
(4-Aminophenyl)acetonitrile 99%
Catalog # ACRO103210100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Yellow to brown Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    41 to 46 °C
  • GC:
    ≥98.5 %

Specification Test Results

About this item

 :
 :132.17 g/mol
Melting Pt:42…46 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:222-587-4
UN:3439
ADR:6.1,III
 :GHS07
CAS Number:3544-25-0
Formula:C₈H₈N₂
MDL Number:MFCD00007912