Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Eschka's mixture analytical reagent, for the determination of sulphur in coal, Supelco®
  1.03162.0250
 :  Merck
 :  
 
Eschka's mixture analytical reagent, for the determination of sulphur in coal, Supelco®
  1.03162.0250
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

 

 

  • Solubility:
    80 g/l (20 °C)
  • Bulk Density:
    300 kg/m3
  • pH:
    12 (50 g/l, H2O, 20 °C)

 

 

 :
 :M-Clarity™ quality level = MQ100
 :GHS07
 :Ambient
 :8007-09-8