


Boron trifluoride tetrahydrofuran complex (45.5% BF₃)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:462-34-0
:
Formula:C₄H₈BF₃O
MDL Number:MFCD00040372
Boiling Pt:180 °C
Melting Pt:8…9 °C
Density:1.268
Flash Pt:92 °C (197 °F)
Storage Temperature:Ambient
EINECS:207-325-9
ADR:8,II
:139.91 g/mol
UN:3265
:GHS08
:GHS05
:GHS07
Related Information
Frequently Bought Together