Order Entry
Czech Republic
ContactUsLinkComponent
o-Dichlorobenzene : Phenol mixture (50:50) (w/w) for synthesis, for measuring the viscosity acc. to DIN 53728, Sigma-Aldrich®
Toxic
Corrosive
Health
Environment
o-Dichlorobenzene : Phenol mixture (50:50) (w/w) for synthesis, for measuring the viscosity acc. to DIN 53728, Sigma-Aldrich®
Catalog # 8.14765.2500
Supplier:  Merck
CAS Number:  
Toxic
Corrosive
Health
Environment
o-Dichlorobenzene : Phenol mixture (50:50) (w/w) for synthesis, for measuring the viscosity acc. to DIN 53728, Sigma-Aldrich®
Catalog # 8.14765.2500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay of phenole (GC, weight%):
    48.0 - 52.0 %
  • Water (K. F.):
    ≤ 200 ppm
  • Assay of dichloro benzene (GC, weight%):
    48.0 - 52.0 %
  • Colourless liquid:
    passes test

Specification Test Results

About this item

 :
ADR:6.1,II
 :M-Clarity™ quality level = MQ200
UN:2810
 :GHS06
 :GHS05
 :GHS08
 :GHS09
Storage Temperature:Ambient