Order Entry
Czech Republic
ContactUsLinkComponent
 
 
2-Fluoroadenosine ≥96.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
 
2-Fluoroadenosine ≥96.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 96.0 area%
  • Purity (Nonaqueous Titration):
    min. 96.0 %
  • Specific rotation [a]20/D:
    -67.0 to -73.0 deg (C=0.1, EtOH)

 

Frequently Bought Together