

cis,cis-1,5-Cyclooctadiene ≥99% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1552-12-1
:
Formula:C₈H₁₂
MDL Number:MFCD00001752
:108.18 g/mol
Boiling Pt:151…152 °C
Melting Pt:–70 °C
Density:0.880
Flash Pt:32 °C (89 °F)
UN:2520
ADR:3,III
:Stabilised with IRGANOX 1076
EINECS:216-291-4
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together