
2-Chlorophenyl isothiocyanate ≥95.0% (by GC)
TCIAI0534-25G
: TCI
:
- Typ balení:Glass bottle
- Bal.:25 g
- Sekvence:3308
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
:
:262°C
:1,29 g/cm³ (20 °C)
:2810
:6.1,III
:GHS06
:2740-81-0
:C₇H₄ClNS
:MFCD00004801