ContactUsLinkComponent
Cotton dispensing jars, with lid
Cotton dispensing jars, with lid
Catalog # HECH44295010
undefined
Cotton dispensing jars, with lid
Catalog # HECH44295010
Specifications
  • Ext.Ø:
    100 mm
  • Height:
    100 mm
  • Pk:
    1
Specifications
About this item
Jars for cotton dispensing, made of glass with stainless steel lid.
  • Sterilisable (without cotton)
  • Good chemical resistance