ContactUsLinkComponent
Test tubes,with rim
  HECH42771015
undefined
Test tubes,with rim
  HECH42771015
  • Ext.Ø:
    7,8 mm
  • Wall thickness:
    0,45 mm
  • Length:
    70 mm
  • Pk:
    100
FIOLAX®, neutral glass.
  • Very good chemical resistance
  • Relatively resistant against rapid temperature changes and local heating
Thin walled test tubes for a wide variety of applications.