Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
 
n-Pentane ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
 :  Merck
 
undefined
 
 
 
 
n-Pentane ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
 :  Merck
 

 

  • Solubility:
    0.04 g/L (25 °C)
  • Melting Point:
    -130 °C
  • Molar Mass:
    72.15 g/mol
  • Boiling Point:
    36 °C (1013 hPa)
  • Vapor Pressure:
    570 hPa (20 °C)
  • Refractive Index:
    1.358
  • Explosion Limit:
    1.4 - 8%(V)
  • Density:
    0.63 g/cm³ (20 °C)
  • pH:
    (H₂O) no data available
  • Ignition Point:
    285 °C
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

 

Frequently Bought Together