2-(Hydroxymethyl)-12-crown-4, Sigma-Aldrich®
Catalog # SIAL382655-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:206.24 g/mol
:M-Clarity™ quality level = MQ200
EINECS:278-224-5
CAS Number:75507-26-5
Formula:C₉H₁₈O₅
MDL Number:MFCD00067524